3-Bromo-4-propoxyphenylacetic acid
Dodavatel: Apollo Scientific
APOSOR471557-1GEA
28700
CZK
APOSOR471557-1G
3-Bromo-4-propoxyphenylacetic acid
3-Bromo-4-propoxyphenylacetic acid
Vzorec:
C₁₁H₁₃BrO₃ MW: 273,123 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26595126 CAS číslo: 1511318-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...