Tisk…

2,3-Diamino-6-chloro-benzonitrile

Dodavatel: Apollo Scientific

APOSOR471585-1GEA 2660 CZK
APOSOR471585-1G
2,3-Diamino-6-chloro-benzonitrile
2,3-Diamino-6-chloro-benzonitrile
Vzorec: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20723943
CAS číslo: 548457-80-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR