Tisk…

1,3-Dibromo-5-isopropoxybenzene 97%

Dodavatel: Apollo Scientific

APOSOR471673-5GEA 4240 CZK
APOSOR471673-5G
1,3-Dibromo-5-isopropoxybenzene 97%
1,3-Dibromo-5-isopropoxybenzene
Vzorec: C₉H₁₀Br₂O
MW: 293,983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547881
CAS číslo: 1112210-82-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR