3-Amino-5-cyanobenzoic acid ≥97%
Dodavatel: Apollo Scientific
APOSOR47859-250MGEA
7050
CZK
APOSOR47859-250MG
3-Amino-5-cyanobenzoic acid ≥97%
3-Amino-5-cyanobenzoic acid
Vzorec:
C₈H₆N₂O₂ MW: 162,145 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09263225 CAS číslo: 1000341-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...