Tisk…

Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR480011-1GEA 50260 CZK
APOSOR480011-1G
Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2-ethoxyethyl)pyrazole-3-carboxylate
Vzorec: C₁₀H₁₆ClN₃O₃
MW: 261,705 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135177
CAS číslo: 1427010-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR