Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Dodavatel: Apollo Scientific
APOSOR480024-1GEA
39020
CZK
APOSOR480024-1G
Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2,2-dimethylpropyl)pyrazole-3-carboxylate
Vzorec:
C₁₁H₁₉N₃O₂ MW: 225,287 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23135190 CAS číslo: 1427023-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...