Tisk…

Ethyl 5-amino-1-tetrahydrofuran-3-yl-pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR480038-1GEA 50590 CZK
APOSOR480038-1G
Ethyl 5-amino-1-tetrahydrofuran-3-yl-pyrazole-3-carboxylate
Ethyl 5-amino-1-tetrahydrofuran-3-yl-pyrazole-3-carboxylate
Vzorec: C₁₀H₁₅N₃O₃
MW: 225,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135209
CAS číslo: 1427020-34-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR