Tisk…

Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR480042-1GEA 25300 CZK
APOSOR480042-1G
Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-phenyl-pyrazole-3-carboxylate
Vzorec: C₁₂H₁₂IN₃O₂
MW: 357,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135213
CAS číslo: 1427011-93-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR