Tisk…

Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR480073-1GEA 19480 CZK
APOSOR480073-1G
Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate
Vzorec: C₁₆H₁₅N₃O₂
MW: 281,309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07388442
CAS číslo: 1427011-96-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR