Tisk…

Ethyl 5-amino-1-(8-quinolyl)pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

APOSOR480078-1GEA 50260 CZK
APOSOR480078-1G
Ethyl 5-amino-1-(8-quinolyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(8-quinolyl)pyrazole-3-carboxylate
Vzorec: C₁₅H₁₄N₄O₂
MW: 282,297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135168
CAS číslo: 1427020-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR