1-(Pyridin-3-yl)cyclopropanamine dihydrochloride
Dodavatel: Apollo Scientific
APOSOR480082-1GEA
34060
CZK
APOSOR480082-1G
1-(Pyridin-3-yl)cyclopropanamine dihydrochloride
1-(Pyridin-3-yl)cyclopropanamine dihydrochloride
Vzorec:
C₈H₁₀N₂·2ClH MW: 207,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08752627 CAS číslo: 1187932-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...