1-(2-Chloro-4-pyridyl)cyclopropanamine hydrochloride
Dodavatel: Apollo Scientific
APOSOR480084-1GEA
64800
CZK
APOSOR480084-1G
1-(2-Chloro-4-pyridyl)cyclopropanamine hydrochloride
1-(2-Chloro-4-pyridyl)cyclopropanamine hydrochloride
Vzorec:
C₈H₉ClN₂·ClH MW: 205,084 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30496518 CAS číslo: 2149601-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...