Tisk…

1-(5-Chloro-2-pyridyl)cyclopropanamine hydrochloride

Dodavatel: Apollo Scientific

APOSOR480085-1GEA 64800 CZK
APOSOR480085-1G
1-(5-Chloro-2-pyridyl)cyclopropanamine hydrochloride
1-(5-Chloro-2-pyridyl)cyclopropanamine hydrochloride
Vzorec: C₈H₉ClN₂·ClH
MW: 205,084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30496521
CAS číslo: 1384264-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR