Tisk…

1-Thiazol-4-ylcyclopropanecarboxylic acid

Dodavatel: Apollo Scientific

APOSOR480111-1GEA 47280 CZK
APOSOR480111-1G
1-Thiazol-4-ylcyclopropanecarboxylic acid
1-Thiazol-4-ylcyclopropanecarboxylic acid
Vzorec: C₇H₇NO₂S
MW: 169,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30496514
CAS číslo: 1542332-02-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR