1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
Dodavatel: Apollo Scientific
APOSOR480161-1GEA
129270
CZK
APOSOR480161-1G
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
Vzorec:
C₁₄H₁₀BrN MW: 272,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30496523 CAS číslo: 2113847-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...