Tisk…

1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine

Dodavatel: Apollo Scientific

APOSOR480165-1GEA 129270 CZK
APOSOR480165-1G
1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine
1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine
Vzorec: C₁₃H₁₂BrN
MW: 262,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30496535
CAS číslo: 1780001-29-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR