Tisk…

DL-threo-3-Phenylserine hydrate 98%

Dodavatel: Apollo Scientific

APOSOR480205-5GEA 2990 CZK
APOSOR480205-5G
DL-threo-3-Phenylserine hydrate 98%
DL-threo-3-Phenylserine hydrate
Vzorec: C₉H₁₁NO₃·H₂O
MW: 199,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863724
CAS číslo: 207605-47-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR