Tisk…

(2S)-2-Aminohept-6-enoic acid 95%

Dodavatel: Apollo Scientific

APOSOR480412-500MGEA 21760 CZK
APOSOR480412-500MG
(2S)-2-Aminohept-6-enoic acid 95%
(2S)-2-Aminohept-6-enoic acid
Vzorec: C₇H₁₃NO₂
MW: 143,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200653
CAS číslo: 166734-64-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR