Tisk…

(5S)-5-Aminopiperidin-2-one hydrochloride 97%

Dodavatel: Apollo Scientific

Varování

APOSOR480824-1GEA 18650 CZK
APOSOR480824-1G
(5S)-5-Aminopiperidin-2-one hydrochloride 97%
(5S)-5-Aminopiperidin-2-one hydrochloride
Vzorec: C₅H₁₀N₂O·ClH
MW: 150,607 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094719
CAS číslo: 672883-95-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR