Tisk…

3,5-Diamino-2-hydroxybenzoic acid 95%

Dodavatel: Apollo Scientific

APOSOR480869-1GEA 6050 CZK
APOSOR480869-1G
3,5-Diamino-2-hydroxybenzoic acid 95%
3,5-Diamino-2-hydroxybenzoic acid
Vzorec: C₇H₈N₂O₃
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753462
CAS číslo: 112725-89-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR