Tisk…

2H-1,4-Benzoxazine-3(4H)-thione

Dodavatel: Apollo Scientific

APOSOR4833-1GEA 5820 CZK
APOSOR4833-1G
2H-1,4-Benzoxazine-3(4H)-thione
2H-1,4-Benzoxazine-3(4H)-thione
Vzorec: C₈H₇NOS
MW: 165,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269330
CAS číslo: 14183-51-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR