Tisk…

2-Amino-3-chloro-5-methylphenol 97%

Dodavatel: Apollo Scientific
Varování

APOSOR500003-250MGEA 3540 CZK
APOSOR500003-250MG
2-Amino-3-chloro-5-methylphenol 97%
2-Amino-3-chloro-5-methylphenol
Vzorec: C₇H₈ClNO
MW: 157,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28551485
CAS číslo: 1780449-40-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR