Tisk…

2-Bromo-4-methylnicotinonitrile 95+%

Dodavatel: Apollo Scientific
Varování

APOSOR500018-250MGEA 2700 CZK
APOSOR500018-250MG
2-Bromo-4-methylnicotinonitrile 95+%
2-Bromo-4-methylnicotinonitrile
Vzorec: C₇H₅BrN₂
MW: 197,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834634
CAS číslo: 65996-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR