Tisk…

2-Methylimidazo[1,2-a]pyridin-6-amine ≥97%

Dodavatel: Apollo Scientific

APOSOR510002-250MGEA 6720 CZK
APOSOR510002-250MG
2-Methylimidazo[1,2-a]pyridin-6-amine ≥97%
2-Methylimidazo[1,2-a]pyridin-6-amine
Vzorec: C₈H₉N₃
MW: 147,177 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991758
CAS číslo: 860258-05-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR