Tisk…

6-Chloro-2,3-dihydro-1H-inden-1-amine ≥97%

Dodavatel: Apollo Scientific

APOSOR510012-250MGEA 5060 CZK
APOSOR510012-250MG
6-Chloro-2,3-dihydro-1H-inden-1-amine ≥97%
6-Chloro-2,3-dihydro-1H-inden-1-amine
Vzorec: C₉H₁₀ClN
MW: 167,635 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07373938
CAS číslo: 67120-38-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR