Tisk…

3-Bromobenzo[b]thiophen-5-amine ≥98%

Dodavatel: Apollo Scientific

APOSOR510038-250MGEA 6390 CZK
APOSOR510038-250MG
3-Bromobenzo[b]thiophen-5-amine ≥98%
3-Bromobenzo[b]thiophen-5-amine
Vzorec: C₈H₆BrNS
MW: 228,109 g/mol
Storage Temperature: Refrigerator
CAS číslo: 98434-36-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR