Tisk…

5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine ≥97%

Dodavatel: Apollo Scientific

APOSOR510042-250MGEA 8340 CZK
APOSOR510042-250MG
5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine ≥97%
5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine
Vzorec: C₇H₄BrClN₂
MW: 231,477 g/mol
Storage Temperature: Refrigerator
CAS číslo: 1612287-20-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR