7-Bromo-4-methoxyquinazoline ≥98%
Dodavatel: Apollo Scientific
APOSOR510047-250MGEA
5100
CZK
APOSOR510047-250MG
7-Bromo-4-methoxyquinazoline ≥98%
7-Bromo-4-methoxyquinazoline
Vzorec:
C₉H₇BrN₂O MW: 239,069 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30834447 CAS číslo: 2140305-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...