Tisk…

5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde ≥95%

Dodavatel: Apollo Scientific

APOSOR510055-250MGEA 8930 CZK
APOSOR510055-250MG
5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde ≥95%
5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde
Vzorec: C₇H₉N₃O
MW: 151,166 g/mol
Storage Temperature: Refrigerator
CAS číslo: 877133-27-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR