Tisk…

5-Iodo-1-methyluracil ≥96%

Dodavatel: Apollo Scientific

APOSOR510075-250MGEA 9200 CZK
APOSOR510075-250MG
5-Iodo-1-methyluracil ≥96%
5-Iodo-1-methyluracil
Vzorec: C₅H₅IN₂O₂
MW: 252,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22578662
CAS číslo: 45774-47-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR