Tisk…

2-Iodo-4-methoxy-1-(methylsulphonyl)benzene ≥97%

Dodavatel: Apollo Scientific

APOSOR510081-250MGEA 2410 CZK
APOSOR510081-250MG
2-Iodo-4-methoxy-1-(methylsulphonyl)benzene ≥97%
2-Iodo-4-methoxy-1-(methylsulphonyl)benzene
Vzorec: C₈H₉IO₃S
MW: 312,125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30181850
CAS číslo: 1965309-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR