Tisk…

Methyl 5-amino-1H-imidazole-4-carboxylate ≥97%

Dodavatel: Apollo Scientific

APOSOR510085-250MGEA 2280 CZK
APOSOR510085-250MG
Methyl 5-amino-1H-imidazole-4-carboxylate ≥97%
Methyl 5-amino-1H-imidazole-4-carboxylate
Vzorec: C₅H₇N₃O₂
MW: 141,128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756040
CAS číslo: 4919-00-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR