Tisk…

4-Bromo-5-nitro-1H-indole ≥97%

Dodavatel: Apollo Scientific

APOSOR510103-250MGEA 9460 CZK
APOSOR510103-250MG
4-Bromo-5-nitro-1H-indole ≥97%
4-Bromo-5-nitro-1H-indole
Vzorec: C₈H₅BrN₂O₂
MW: 241,042 g/mol
Storage Temperature: Refrigerator
CAS číslo: 1934562-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR