Tisk…

6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid ≥97%

Dodavatel: Apollo Scientific

APOSOR510105-250MGEA 4070 CZK
APOSOR510105-250MG
6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid ≥97%
6-Bromoimidazo[1,2-a]pyrimidine-2-carboxylic acid
Vzorec: C₇H₄BrN₃O₂
MW: 242,03 g/mol
Storage Temperature: Refrigerator
CAS číslo: 907945-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR