Tisk…

2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%

Dodavatel: Apollo Scientific

APOSOR510128-100MGEA 13430 CZK
APOSOR510128-100MG
2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine
Vzorec: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13175869
CAS číslo: 7546-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR