Tisk…

7-Chloroisoquinolin-8-amine ≥98%

Dodavatel: Apollo Scientific

APOSOR510144-100MGEA 12730 CZK
APOSOR510144-100MG
7-Chloroisoquinolin-8-amine ≥98%
7-Chloroisoquinolin-8-amine
Vzorec: C₉H₇ClN₂
MW: 178,618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18823041
CAS číslo: 55766-90-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR