4-Iodo-2-methyl-1-nitrobenzene ≥98%
Dodavatel: Apollo Scientific
APOSOR510170-100MGEA
7970
CZK
APOSOR510170-100MG
4-Iodo-2-methyl-1-nitrobenzene ≥98%
4-Iodo-2-methyl-1-nitrobenzene
Vzorec:
C₇H₆INO₂ MW: 263,033 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234827 CAS číslo: 52415-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...