Tisk…

rac-AntPhos ≥98%

Dodavatel: Apollo Scientific

APOSOR510214-1GEA 10490 CZK
APOSOR510214-1G
rac-AntPhos ≥98%
rac-AntPhos
Vzorec: C₂₅H₂₃OP
MW: 370,423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30748199
CAS číslo: 1268693-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR