Tisk…

4-Amino-2-chloro-3-methylbenzonitrile hydrochloride ≥98%

Dodavatel: Apollo Scientific

APOSOR510226-250MGEA 10350 CZK
APOSOR510226-250MG
4-Amino-2-chloro-3-methylbenzonitrile hydrochloride ≥98%
4-Amino-2-chloro-3-methylbenzonitrile hydrochloride
Vzorec: C₈H₇ClN₂·ClH
MW: 203,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858333
CAS číslo: 864361-78-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR