Tisk…

(R)-BI-DIME ≥98%

Dodavatel: Apollo Scientific

APOSOR510253-100MGEA 7610 CZK
APOSOR510253-100MG
(R)-BI-DIME ≥98%
(R)-BI-DIME
Vzorec: C₁₉H₂₃O₃P
MW: 330,358 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31630704
CAS číslo: 1338454-03-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR