2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
Dodavatel: Apollo Scientific
APOSOR5155T-5MGEA
9130
CZK
APOSOR5155T-5MG
2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
Vzorec:
C₁₄H₂₅NO₁₀ MW: 367,349 g/mol Bod tání: 216…218 °C (rozkl.) Storage Temperature: Freezer |
MDL Number:
MFCD00153897 CAS číslo: 76211-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...