Tisk…

Methylterephthalonitrile

Dodavatel: Apollo Scientific
Varování

APOSOR51883-5GEA 2820 CZK
APOSOR51883-5G
Methylterephthalonitrile
Methylterephthalonitrile
Vzorec: C₉H₆N₂
MW: 142,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119249
CAS číslo: 55984-93-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR