Tisk…

5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole

Dodavatel: Apollo Scientific

APOSOR52113-1GEA 15440 CZK
APOSOR52113-1G
5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole
Vzorec: C₇H₅ClN₂S₂
MW: 216,711 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20257108
CAS číslo: 1338218-71-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR