Tisk…

Cyclotrimethylenedimethylsilane

Dodavatel: Apollo Scientific

Nebezpečí

APOSOR52271-5GEA 9490 CZK
APOSOR52271-5G
Cyclotrimethylenedimethylsilane
Cyclotrimethylenedimethylsilane
Vzorec: C₅H₁₂Si
MW: 100,234 g/mol
Hustota: 0,777 g/cm³ (25 °C)
Bod vzplanutí: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00039674
CAS číslo: 2295-12-7
EINECS: 218-939-1
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR