Tisk…

Bicyclo[3.2.1]octan-3-one 95+%

Dodavatel: Apollo Scientific
Varování

APOSOR52275-250MGEA 14520 CZK
APOSOR52275-250MG
Bicyclo[3.2.1]octan-3-one 95+%
Bicyclo[3.2.1]octan-3-one
Vzorec: C₈H₁₂O
MW: 124,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19300751
CAS číslo: 14252-05-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR