Tisk…

[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride

Dodavatel: Apollo Scientific

Varování

APOSOR52550-1GEA 25100 CZK
APOSOR52550-1G
[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride
[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride
Vzorec: C₇H₁₀N₂S·ClH
MW: 190,694 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16990682
CAS číslo: 60481-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR