Tisk…

5-Bromo-2-hydroxy-4-methoxybenzaldehyde

Dodavatel: Apollo Scientific

APOSOR52694-1GEA 3410 CZK
APOSOR52694-1G
5-Bromo-2-hydroxy-4-methoxybenzaldehyde
5-Bromo-2-hydroxy-4-methoxybenzaldehyde
Vzorec: C₈H₇BrO₃
MW: 231,043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00839421
CAS číslo: 57543-36-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR