Tisk…

2-Bromo-3-(1-piperidyl)pyridine 95%

Dodavatel: Apollo Scientific

APOSOR52757-100MGEA 19680 CZK
APOSOR52757-100MG
2-Bromo-3-(1-piperidyl)pyridine 95%
2-Bromo-3-(1-piperidyl)pyridine
Vzorec: C₁₀H₁₃BrN₂
MW: 241,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26405402
CAS číslo: 1563532-94-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR