Tisk…

3-Morpholinopyridin-2-amine 95%

Dodavatel: Apollo Scientific

APOSOR52777-250MGEA 17390 CZK
APOSOR52777-250MG
3-Morpholinopyridin-2-amine 95%
3-Morpholinopyridin-2-amine
Vzorec: C₉H₁₃N₃O
MW: 179,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848423
CAS číslo: 1036645-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR