Tisk…

tert-Butyl N-(2-aminophenyl)-N-methylcarbamate

Dodavatel: Apollo Scientific

APOSOR52955-1GEA 28900 CZK
APOSOR52955-1G
tert-Butyl N-(2-aminophenyl)-N-methylcarbamate
tert-Butyl N-(2-aminophenyl)-N-methylcarbamate
Vzorec: C₁₂H₁₈N₂O₂
MW: 222,284 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05663971
CAS číslo: 885270-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR